accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 47173-80-8

IUPAC Name: (2R)-3-(benzyloxy)-2-{[(tert-butoxy)carbonyl]amino}propanoate
Synonyms: (2R)-3-(benzyloxy)-2-[(tert-butoxycarbonyl)amino]propanoate
Molecular Weight (g/mol): 294.33
Molecular Formula: C15H20NO5
InChi Key: DMBKPDOAQVGTST-GFCCVEGCSA-M
SMILES: CC(C)(C)OC(=O)N[C@H](COCC1=CC=CC=C1)C([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)