accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 471929-86-9

IUPAC Name: 4-[(piperidin-1-yl)methyl]benzaldehyde
Synonyms: 4-(piperidin-1-ylmethyl)benzaldehyde
Molecular Weight (g/mol): 203.29
Molecular Formula: C13H17NO
InChi Key: WWBOSCKXPMBVLG-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(CN2CCCCC2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results