accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4721-17-9

IUPAC Name: 1,3-diethyl 2-({[4-(4-methylphenyl)-1,3-thiazol-2-yl]amino}methylidene)propanedioate
Synonyms: 1,3-diethyl 2-({[4-(4-methylphenyl)-1,3-thiazol-2-yl]amino}methylidene)propanedioate
Molecular Weight (g/mol): 360.43
Molecular Formula: C18H20N2O4S
InChi Key: GSMRMOCCGZENCI-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=CNC1=NC(=CS1)C1=CC=C(C)C=C1)C(=O)OCC
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)