accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4721-24-8

IUPAC Name: hexylphosphonate
Synonyms: hexylphosphonate
Molecular Weight (g/mol): 164.14
Molecular Formula: C6H13O3P
InChi Key: GJWAEWLHSDGBGG-UHFFFAOYSA-L
SMILES: CCCCCCP([O-])([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)