CAS RN 4721-98-6
CAS RN 4721-98-6
IUPAC Name:
6,7-dimethoxy-1-methyl-3,4-dihydroisoquinoline
Synonyms:
6,7-dimethoxy-1-methyl-3,4-dihydroisoquinoline
Molecular Weight (g/mol):
205.26
Molecular Formula:
C12H15NO2
InChi Key:
VASUQTGZAPZKFK-UHFFFAOYSA-N
SMILES:
COC1=CC2=C(C=C1OC)C(C)=NCC2
1
–
1
of
1
results
1
Thermo Scientific Chemicals 1-Methyl-6,7-dimethoxy-3,4-dihydroisoquinoline, 99+%
CAS: 4721-98-6 Molecular Formula: C12H15NO2 Molecular Weight (g/mol): 205.26 MDL Number: MFCD00040276 InChI Key: VASUQTGZAPZKFK-UHFFFAOYSA-N Synonym: 1-methyl-6,7-dimethoxy-3,4-dihydroisoquinoline,isoquinoline, 3,4-dihydro-6,7-dimethoxy-1-methyl,6,7-dimethoxy-1-methyl-3,4-dihydro-isoquinoline,dehydrosalsolidine,maybridge1_001068,3,4-dihydro-6,7-dimethoxy-1-methylisoquinoline,vasuqtgzapzkfk-uhfffaoysa,1-methyl-3,4-dihydro-6,7-dimethoxyisoquinoline,1-methyl-6?7-dimethoxy-3?4-dihydroisoquinoline,# PubChem CID: 22652 IUPAC Name: 6,7-dimethoxy-1-methyl-3,4-dihydroisoquinoline SMILES: CC1=NCCC2=CC(=C(C=C12)OC)OC