CAS RN 473-29-0
IUPAC Name:
N,N-dichlorobenzenesulfonamide
Synonyms:
N,N-dichlorobenzenesulfonamide
Molecular Weight (g/mol):
226.07
Molecular Formula:
C6H5Cl2NO2S
InChi Key:
PJBJJXCZRAHMCK-UHFFFAOYSA-N
SMILES:
ClN(Cl)S(=O)(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
Dichloramine B 95.0+%, TCI America™
CAS: 473-29-0 Molecular Formula: C6H5Cl2NO2S Molecular Weight (g/mol): 226.07 MDL Number: MFCD00025025 InChI Key: PJBJJXCZRAHMCK-UHFFFAOYSA-N Synonym: Benzenesulfonedichloramide PubChem CID: 68055 IUPAC Name: N,N-dichlorobenzenesulfonamide SMILES: ClN(Cl)S(=O)(=O)C1=CC=CC=C1