accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 473-81-4

IUPAC Name: potassium 2,3-dihydroxypropanoate
Synonyms: glyceric acid
Molecular Weight (g/mol): 144.17
Molecular Formula: C3H5KO4
InChi Key: SYGDCNJESIQXKS-UHFFFAOYNA-M
SMILES: [K+].OCC(O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results