accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4730-22-7

IUPAC Name: 6-methylheptan-2-ol
Synonyms: 6-methyl-2-heptanol
Molecular Weight (g/mol): 130.23
Molecular Formula: C8H18O
InChi Key: FCOUHTHQYOMLJT-UHFFFAOYNA-N
SMILES: CC(C)CCCC(C)O
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)