accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4737-50-2

IUPAC Name: pentylboronic acid
Synonyms: pentylboronic acid
Molecular Weight (g/mol): 115.97
Molecular Formula: C5H13BO2
InChi Key: ABWPXVJNCQKYDR-UHFFFAOYSA-N
SMILES: CCCCCB(O)O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results