accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 473717-08-7

IUPAC Name: N9,N10-bis(naphthalen-2-yl)-N9,N10-diphenylanthracene-9,10-diamine
Synonyms: N9,N10-bis(naphthalen-2-yl)-N9,N10-diphenylanthracene-9,10-diamine
Molecular Weight (g/mol): 612.78
Molecular Formula: C46H32N2
InChi Key: LIBLRJBCMMHCHS-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)N(C1=CC=C2C=CC=CC2=C1)C1=C2C=CC=CC2=C(N(C2=CC=CC=C2)C2=CC=C3C=CC=CC3=C2)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results