accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 474024-36-7

IUPAC Name: 2-[2-bromo-4-(trifluoromethyl)phenyl]acetonitrile
Synonyms: 2-[2-bromo-4-(trifluoromethyl)phenyl]acetonitrile
Molecular Weight (g/mol): 264.05
Molecular Formula: C9H5BrF3N
InChi Key: RYUZTTSDBHOIAF-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Br)=C(CC#N)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results