accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOONN

CAS RN 4744-50-7

IUPAC Name: 5H,7H-furo[3,4-b]pyrazine-5,7-dione
Synonyms: furo[3,4-b]pyrazine-5,7-dione
Molecular Weight (g/mol): 150.09
Molecular Formula: C6H2N2O3
InChi Key: AWJWCTOOIBYHON-UHFFFAOYSA-N
SMILES: O=C1OC(=O)C2=NC=CN=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results