accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 47458-32-2

IUPAC Name: 3-octadecyloxolane-2,5-dione
Synonyms: 3-octadecyloxolane-2,5-dione
Molecular Weight (g/mol): 352.56
Molecular Formula: C22H40O3
InChi Key: ZJFCVUTYZHUNSW-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCCCCCC1CC(=O)OC1=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results