accessibility menu, dialog, popup

UserName

Viewing profile as user:
Br

CAS RN 474688-73-8

IUPAC Name: 9-bromo-10-(naphthalen-2-yl)anthracene
Synonyms: 9-bromo-10-(naphthalen-2-yl)anthracene
Molecular Weight (g/mol): 383.29
Molecular Formula: C24H15Br
InChi Key: FKIFDWYMWOJKTQ-UHFFFAOYSA-N
SMILES: BrC1=C2C=CC=CC2=C(C2=CC3=CC=CC=C3C=C2)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)