accessibility menu, dialog, popup

UserName

Viewing profile as user:
Br

CAS RN 474688-74-9

IUPAC Name: 9-bromo-10-{5-phenyl-[1,1'-biphenyl]-3-yl}anthracene
Synonyms: 9-bromo-10-{5-phenyl-[1,1'-biphenyl]-3-yl}anthracene
Molecular Weight (g/mol): 485.42
Molecular Formula: C32H21Br
InChi Key: BQYDXENQXPPQLP-UHFFFAOYSA-N
SMILES: BrC1=C2C=CC=CC2=C(C2=CC(=CC(=C2)C2=CC=CC=C2)C2=CC=CC=C2)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)