accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4747-71-1

IUPAC Name: 14,15-dimethyl-10-phenyl-2,11-diazatricyclo[10.4.0.0⁴,⁹]hexadeca-1(12),4,6,8,10,13,15-heptaen-3-one
Synonyms: 14,15-dimethyl-10-phenyl-2,11-diazatricyclo[10.4.0.0⁴,⁹]hexadeca-1(12),4,6,8,10,13,15-heptaen-3-one
Molecular Weight (g/mol): 326.4
Molecular Formula: C22H18N2O
InChi Key: UHJCHZGAXRAPFM-UHFFFAOYSA-N
SMILES: CC1=CC2=C(C=C1C)N=C(C1=CC=CC=C1)C1=CC=CC=C1C(=O)N2
Boiling Point 
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)