accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4750-28-1

IUPAC Name: N-(4-chlorophenyl)benzenesulfonamide
Synonyms: N-(4-chlorophenyl)benzenesulfonamide
Molecular Weight (g/mol): 267.73
Molecular Formula: C12H10ClNO2S
InChi Key: ANRCRHLXUCJAKV-UHFFFAOYSA-N
SMILES: ClC1=CC=C(NS(=O)(=O)C2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results