accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4755-77-5

IUPAC Name: ethyl oxalochloridate
Synonyms: ethyl chloroglyoxylate
Molecular Weight (g/mol): 136.53
Molecular Formula: C4H5ClO3
InChi Key: OWZFULPEVHKEKS-UHFFFAOYSA-N
SMILES: CCOC(=O)C(Cl)=O
Boiling Point 
  • (3)
  • (9)
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (6)
  • (3)
  • (9)
  • (3)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (3)
  • (2)
  • (6)
  • (3)
  • (6)
  • (6)