accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOClCH3O

CAS RN 4755-81-1

IUPAC Name: methyl 2-chloro-3-oxobutanoate
Synonyms: methyl 2-chloro-3-oxobutanoate
Molecular Weight (g/mol): 150.56
Molecular Formula: C5H7ClO3
InChi Key: GYQRIAVRKLRQKP-UHFFFAOYNA-N
SMILES: COC(=O)C(Cl)C(C)=O
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)