accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4760-34-3

IUPAC Name: N1-methylbenzene-1,2-diamine
Synonyms: N1-methylbenzene-1,2-diamine
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: RPKCLSMBVQLWIN-UHFFFAOYSA-N
SMILES: CNC1=CC=CC=C1N
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)