accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4766-57-8

IUPAC Name: tetrabutyl silicate
Synonyms: tetrabutyl silicate
Molecular Weight (g/mol): 320.55
Molecular Formula: C16H36O4Si
InChi Key: UQMOLLPKNHFRAC-UHFFFAOYSA-N
SMILES: CCCCO[Si](OCCCC)(OCCCC)OCCCC
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)