accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4767-00-4

IUPAC Name: 3-(1,3-benzothiazol-2-ylsulfanyl)propanoic acid
Synonyms: 3-(1,3-benzothiazol-2-ylsulfanyl)propanoic acid
Molecular Weight (g/mol): 239.31
Molecular Formula: C10H9NO2S2
InChi Key: DXSBAOMLHPFLMW-UHFFFAOYSA-N
SMILES: OC(=O)CCSC1=NC2=CC=CC=C2S1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results