accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3COOHN(R)OOOBrOHO

CAS RN 47689-67-8

IUPAC Name: (2R)-3-[4-({[(2-bromophenyl)methoxy]carbonyl}oxy)phenyl]-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Synonyms: (2R)-3-[4-({[(2-bromophenyl)methoxy]carbonyl}oxy)phenyl]-2-[(tert-butoxycarbonyl)amino]propanoic acid
Molecular Weight (g/mol): 494.34
Molecular Formula: C22H24BrNO7
InChi Key: UYWMYJQSTUVRHR-GOSISDBHSA-N
SMILES: CC(C)(C)OC(=O)N[C@H](CC1=CC=C(OC(=O)OCC2=CC=CC=C2Br)C=C1)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)