accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHOOO

CAS RN 477-84-9

IUPAC Name: 3-hydroxy-1-methoxy-9,10-dioxo-9,10-dihydroanthracene-2-carbaldehyde
Synonyms: damnacanthal
Molecular Weight (g/mol): 282.25
Molecular Formula: C16H10O5
InChi Key: IPDMWUNUULAXLU-UHFFFAOYSA-N
SMILES: COC1=C(C=O)C(O)=CC2=C1C(=O)C1=CC=CC=C1C2=O
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results