accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 477-90-7

IUPAC Name: (2S,4R,5S,6S,7R)-5,6,12,14-tetrahydroxy-4-(hydroxymethyl)-13-methoxy-3,8-dioxatricyclo[8.4.0.0²,⁷]tetradeca-1(14),10,12-trien-9-one
Synonyms: bergenin bergenin hydrate
Molecular Weight (g/mol): 328.27
Molecular Formula: C14H16O9
InChi Key: YWJXCIXBAKGUKZ-HJJNZUOJSA-N
SMILES: COC1=C(O)C=C2C(=O)O[C@@H]3[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]3C2=C1O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results