accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4774-14-5

IUPAC Name: 2,6-dichloropyrazine
Synonyms: 2-chloro-6-chloro-pyrazine
Molecular Weight (g/mol): 148.97
Molecular Formula: C4H2Cl2N2
InChi Key: LSEAAPGIZCDEEH-UHFFFAOYSA-N
SMILES: ClC1=CN=CC(Cl)=N1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)