accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 478043-88-8

IUPAC Name: 4-(2-formylphenoxy)benzonitrile
Synonyms: 4-(2-formylphenoxy)benzonitrile
Molecular Weight (g/mol): 223.23
Molecular Formula: C14H9NO2
InChi Key: PHXACBMAMQKNJM-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC=C1OC1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results