accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 479-18-5

IUPAC Name: 7-(2,3-dihydroxypropyl)-1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms: dyphylline diprofilina neutrafillina lunsco brand of dyphylline neutraphylline liactemin diphylline wallace brand of dyphylline savage brand of dyphylline hauck brand of dyphylline
Molecular Weight (g/mol): 254.25
Molecular Formula: C10H14N4O4
InChi Key: KSCFJBIXMNOVSH-UHFFFAOYNA-N
SMILES: CN1C2=C(N(CC(O)CO)C=N2)C(=O)N(C)C1=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Physical Form 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)