CAS RN 479-41-4
CAS RN 479-41-4
IUPAC Name:
(E)-1',2'-dihydro-1H,3H-[2,3'-biindolylidene]-2',3-dione
Synonyms:
indirubin
Molecular Weight (g/mol):
262.27
Molecular Formula:
C16H10N2O2
InChi Key:
CRDNMYFJWFXOCH-BUHFOSPRSA-N
SMILES:
O=C1NC2=CC=CC=C2\C1=C1/NC2=CC=CC=C2C1=O
1
–
1
of
1
results
1
Indirubin 97.0+%, TCI America™
CAS: 479-41-4 Molecular Formula: C16H10N2O2 Molecular Weight (g/mol): 262.27 MDL Number: MFCD00956441,MFCD00221745 InChI Key: CRDNMYFJWFXOCH-BUHFOSPRSA-N Synonym: Indigo Red PubChem CID: 5380743 IUPAC Name: (E)-1',2'-dihydro-1H,3H-[2,3'-biindolylidene]-2',3-dione SMILES: O=C1NC2=CC=CC=C2\C1=C1/NC2=CC=CC=C2C1=O