accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 479-45-8

IUPAC Name: N-methyl-N,2,4,6-tetranitroaniline
Synonyms: tetryl N-methyl-N-picrylnitramine N-picryl-N-methylnitramine (trinitrophenyl)methylnitramine
Molecular Weight (g/mol): 287.14
Molecular Formula: C7H5N5O8
InChi Key: AGUIVNYEYSCPNI-UHFFFAOYSA-N
SMILES: CN(C1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results