accessibility menu, dialog, popup

UserName

Viewing profile as user:
O

CAS RN 479-79-8

IUPAC Name: 11H-benzo[a]fluoren-11-one
Synonyms: benzo[a]fluoren-11-one
Molecular Weight (g/mol): 230.27
Molecular Formula: C17H10O
InChi Key: RNICURKFVSAHLQ-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2C2=CC=C3C=CC=CC3=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results