accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 479-92-5

IUPAC Name: 1,5-dimethyl-2-phenyl-4-(propan-2-yl)-2,3-dihydro-1H-pyrazol-3-one
Synonyms: demex isopropylantipyrin isopropyrine declimed brand of propyphenazone yoshipyrin isopropylantipyrinum propifenazone propifenazona propyphenazone larodon
Molecular Weight (g/mol): 230.31
Molecular Formula: C14H18N2O
InChi Key: PXWLVJLKJGVOKE-UHFFFAOYSA-N
SMILES: CC(C)C1=C(C)N(C)N(C1=O)C1=CC=CC=C1
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results