accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNNOHO

CAS RN 479553-01-0

IUPAC Name: 1H-pyrrolo[2,3-b]pyridine-4-carboxylic acid
Synonyms: 1H-pyrrolo[2,3-b]pyridine-4-carboxylic acid
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: HPPPHDPVSYYWCH-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2C=CNC2=NC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results