accessibility menu, dialog, popup

UserName

Viewing profile as user:
NMgBr

CAS RN 480424-82-6

IUPAC Name: 1-{[3-(bromomagnesio)phenyl]methyl}piperidine
Synonyms: 1-{[3-(bromomagnesio)phenyl]methyl}piperidine
Molecular Weight (g/mol): 278.48
Molecular Formula: C12H16BrMgN
InChi Key: QLVYNMWCYBTNEP-UHFFFAOYSA-M
SMILES: Br[Mg]C1=CC=CC(CN2CCCCC2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results