accessibility menu, dialog, popup

UserName

Viewing profile as user:
NMgBr

CAS RN 480424-83-7

IUPAC Name: magnesium(2+) 4-[(piperidin-1-yl)methyl]benzen-1-ide bromide
Synonyms: magnesium(2+) 4-(piperidin-1-ylmethyl)benzen-1-ide bromide
Molecular Weight (g/mol): 278.48
Molecular Formula: C12H16BrMgN
InChi Key: XDZALNKSOIYDJE-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].C(N1CCCCC1)C1=CC=[C-]C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results