accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHBOH3CH3CH3CCH3O

CAS RN 480424-93-9

IUPAC Name: N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetamide
Synonyms: N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetamide
Molecular Weight (g/mol): 261.13
Molecular Formula: C14H20BNO3
InChi Key: CZFSGYCLOCCASM-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results