accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 480438-58-2

IUPAC Name: (2-ethoxy-4-fluorophenyl)boronic acid
Synonyms: 2-ethoxy-4-fluorophenylboronic acid
Molecular Weight (g/mol): 183.97
Molecular Formula: C8H10BFO3
InChi Key: WCJKVQYVILAKQW-UHFFFAOYSA-N
SMILES: CCOC1=C(C=CC(F)=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results