accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 480438-59-3

IUPAC Name: [4-fluoro-2-(propan-2-yloxy)phenyl]boronic acid
Synonyms: 4-fluoro-2-isopropoxyphenylboronic acid
Molecular Weight (g/mol): 198
Molecular Formula: C9H12BFO3
InChi Key: RSEFJQODCQCGRB-UHFFFAOYSA-N
SMILES: CC(C)OC1=C(C=CC(F)=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results