accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H2NNHOOCH3CH3H3C

CAS RN 480452-05-9

IUPAC Name: tert-butyl N-(3-amino-2-methylpropyl)carbamate
Synonyms: tert-butyl N-(3-amino-2-methylpropyl)carbamate
Molecular Weight (g/mol): 188.27
Molecular Formula: C9H20N2O2
InChi Key: ORDMEFMUSKDHOL-UHFFFAOYNA-N
SMILES: CC(CN)CNC(=O)OC(C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)