accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 480996-05-2

IUPAC Name: {4'-bromo-[1,1'-biphenyl]-4-yl}boronic acid
Synonyms: 4'-bromo-[1,1'-biphenyl]-4-ylboronic acid
Molecular Weight (g/mol): 276.92
Molecular Formula: C12H10BBrO2
InChi Key: HQAGDFJHKPSZHT-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)C1=CC=C(Br)C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results