accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOO

CAS RN 481-39-0

IUPAC Name: 5-hydroxy-1,4-dihydronaphthalene-1,4-dione
Synonyms: walnut extract 5-hydroxy-1,4-naftochinon juglone toxin caswell no. 515AA juglone toxin (substance) 5-hydroxy-alphanaphthaquinone C.I. natural brown 7
Molecular Weight (g/mol): 174.16
Molecular Formula: C10H6O3
InChi Key: KQPYUDDGWXQXHS-UHFFFAOYSA-N
SMILES: OC1=CC=CC2=C1C(=O)C=CC2=O
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (2)
  • (4)
  • (2)
Quantity 
  • (4)
  • (4)
Physical Form 
  • (4)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)
  • (6)
  • (2)
  • (4)
  • (2)
  • (4)
  • (4)
  • (4)
  • (1)
  • (1)