accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NH2

CAS RN 481-91-4

IUPAC Name: [1,1'-binaphthalene]-4,4'-diamine
Synonyms: [1,1'-binaphthalene]-4,4'-diamine
Molecular Weight (g/mol): 284.36
Molecular Formula: C20H16N2
InChi Key: RZPBZEISZUFQSV-UHFFFAOYSA-N
SMILES: NC1=C2C=CC=CC2=C(C=C1)C1=C2C=CC=CC2=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results