accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4813-57-4

IUPAC Name: octadecyl prop-2-enoate
Synonyms: octadecyl prop-2-enoate
Molecular Weight (g/mol): 324.55
Molecular Formula: C21H40O2
InChi Key: FSAJWMJJORKPKS-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCOC(=O)C=C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)