accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2COOO

CAS RN 48145-04-6

IUPAC Name: 2-phenoxyethyl prop-2-enoate
Synonyms: 2-phenoxyethyl prop-2-enoate
Molecular Weight (g/mol): 192.21
Molecular Formula: C11H12O3
InChi Key: RZVINYQDSSQUKO-UHFFFAOYSA-N
SMILES: C=CC(=O)OCCOC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)