accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4815-30-9

IUPAC Name: 2,4-diethyl 5-amino-3-methylthiophene-2,4-dicarboxylate
Synonyms: 2,4-diethyl 5-amino-3-methylthiophene-2,4-dicarboxylate
Molecular Weight (g/mol): 257.3
Molecular Formula: C11H15NO4S
InChi Key: DGVXLHAJVRRLGV-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)C(C(=O)OCC)=C(N)S1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results