accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOHNOHOOHO

CAS RN 4816-80-2

IUPAC Name: 2-(4-methylbenzenesulfonamido)pentanedioic acid
Synonyms: 2-(4-methylbenzenesulfonamido)pentanedioic acid
Molecular Weight (g/mol): 301.31
Molecular Formula: C12H15NO6S
InChi Key: KKOZKXBAPIYWAT-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NC(CCC(O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)