accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH(S)NOOOO

CAS RN 48172-10-7

IUPAC Name: (2S)-4-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)-2-hydroxybutanoate
Synonyms: (2S)-4-(1,3-dioxoisoindol-2-yl)-2-hydroxybutanoate
Molecular Weight (g/mol): 248.22
Molecular Formula: C12H10NO5
InChi Key: YWDXODQRCDEZLN-VIFPVBQESA-M
SMILES: O[C@@H](CCN1C(=O)C2=CC=CC=C2C1=O)C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results