accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COPSOCH3OClBrCl

CAS RN 4824-78-6

IUPAC Name: O-4-bromo-2,5-dichlorophenyl O,O-diethyl phosphorothioate
Synonyms: bromophos ethyl
Molecular Weight (g/mol): 394.04
Molecular Formula: C10H12BrCl2O3PS
InChi Key: KWGUFOITWDSNQY-UHFFFAOYSA-N
SMILES: CCOP(=S)(OCC)OC1=CC(Cl)=C(Br)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results