accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 483-63-6

CH3NOCH3H3C(E)

CAS RN 483-63-6

IUPAC Name: (2E)-N-ethyl-N-(2-methylphenyl)but-2-enamide
Synonyms: crotamiton crotalgin crotamitone 克罗米通 crotaglin
Molecular Weight (g/mol): 203.29
Molecular Formula: C13H17NO
InChi Key: DNTGGZPQPQTDQF-XBXARRHUSA-N
SMILES: CCN(C(=O)\C=C\C)C1=CC=CC=C1C
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results