CAS RN 4832-52-4
IUPAC Name:
{[bis(phenylsulfanyl)methyl]sulfanyl}benzene
Synonyms:
{[bis(phenylsulfanyl)methyl]sulfanyl}benzene
Molecular Weight (g/mol):
340.52
Molecular Formula:
C19H16S3
InChi Key:
YOQHDHPBEXTHCP-UHFFFAOYSA-N
SMILES:
S(C(SC1=CC=CC=C1)SC1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Tris(phenylthio)methane 98.0+%, TCI America™
CAS: 4832-52-4 Molecular Formula: C19H16S3 Molecular Weight (g/mol): 340.52 MDL Number: MFCD00010396 InChI Key: YOQHDHPBEXTHCP-UHFFFAOYSA-N PubChem CID: 615706 IUPAC Name: {[bis(phenylsulfanyl)methyl]sulfanyl}benzene SMILES: S(C(SC1=CC=CC=C1)SC1=CC=CC=C1)C1=CC=CC=C1